Learn More
Greiner Bio-One Standard Tube Holder, Vacuette™
Product Code. 16030540
Change view
Click to view available options
Sterility:
Non-sterile
Unit Size:
Pack of 1200
Product Code. Sterility Unit Size
16030540 Non-sterile Pack of 1200
1 options
Product Code. 16030540 Supplier Greiner Bio-One Supplier No. 450201
  • ONLINE EXCLUSIVE PRICE
    91.25 €
    71.01 € / Pack of 1200
    Save 20.24 €
    22% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Greiner Bio-One™ Vacuette™ Standard Tube Holders have a ridged surface for better grip and a safer blood collection process.
  • For holding standard Vacuette™ tubes
  • Not-sterile plastic construction
  • Ridged design for better grip and safer handing
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Product Type Standard Tube Holder
Sterility Non-sterile