missing translation for 'onlineSavingsMsg'
Learn More

Stannic Chloride, 5-Hydrate, Lump, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15548194 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Pakningsstørrelse:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Pakningsstørrelse
15548194 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15548194 Leverandør Avantor J.T.Baker Leverandørnr. 397101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High quality chemicals for laboratory and specialized industrial use

Tekniske data

CAS 10026-06-9
Physical Form Lump
Quantity 500 g
Molecular Formula Cl4H10O5Sn
MDL Number MFCD00149864
Synonym tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o
InChI Key KHMOASUYFVRATF-UHFFFAOYSA-J
SMILES O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
IUPAC Name tin(4+) pentahydrate tetrachloride
Molecular Weight (g/mol) 350.59
PubChem CID 61442
Formula Weight 350.58
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.04kg
Chemical Name or Material Stannic Chloride, 5-Hydrate
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.