missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride Dihydrate, MP Biomedicals

Product Code. 11466362 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466362 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11466362 Supplier MP Biomedicals Supplier No. 0210294980
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell Culture Reagent

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2 · 2H2O

Specifications

Quantity 100 g
Formula Weight 225.6
Chemical Name or Material Stannous Chloride Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.