missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride, MP Biomedicals™

Product Code. 11416612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
11416612 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 11416612

Brand: MP Biomedicals 0215015790

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Stannous Chloride, MP Biomedicals is used as a reducing agent, as a reagent in analytical chemistry and as a metal catalyst in organic reactions.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 652°C (Lit.)
Melting Point 246°C (Lit.)
CAS 7772-99-8
Physical Form Crystalline Powder
Quantity 500 g
Molecular Formula Cl2Sn
MDL Number MFCD00011241
Synonym Tin (II) Chloride, Tin Dichloride, Tin Protochloride
Solubility Information Soluble in water.
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Sn++]
IUPAC Name λ²-tin(2+) dichloride
Molecular Weight (g/mol) 189.61
PubChem CID 24479
ChEBI CHEBI:78067
Formula Weight 189.6
Percent Purity ∼99%
Density 7.3g/cm3 at 20°C (Lit.)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.