missing translation for 'onlineSavingsMsg'
Learn More

Novacyt - Microgen Bioproducts Staph Control Reagent

Product Code. 17393191 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
7 bottles (5 x 5 mL, 2 x 1 mL)
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17393191 7 bottles (5 x 5 mL, 2 x 1 mL) 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17393191 Supplier Novacyt - Microgen Bioproducts Supplier No. M43CCE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microgen Bioproducts Staph Control Reagent for use with M43CE/M433CE.

  • Detects both protein A and coagulase enabling the specific identification of Staphylococcus aureus.

Specifications

Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
Control Sets M43B Positive Control 2 x 1 mL
CE Marker Yes
Clia Complexity Non-waived
Detectable Analytes Fibrinogen and IgG
Format 7 Bottles (5 x 5 mL, 2 x 1 mL)
Sample Type Bacterial Colonies
Sensitivity 1
Type Agglutination Test
For Use With (Application) Confirmatory Identification of Presumptive Staphylococcus Aureuscolonies From Primary Plate Culture
For Use With (Equipment) Test Slide, Sterile Loop
Includes 100 tests
No. of Reactions 1/Test
Organism Group Monera
Packaging Type Carton with Insert vial box
Quantity 7 bottles (5 x 5 mL, 2 x 1 mL)
Reagent Type Staph Latex Reagent 5 mL
Species Latex coated with human fibrinogen
Specificity 1
Target Organism Class Bacteria
Test Time 2 minutes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.