missing translation for 'onlineSavingsMsg'
Learn More

Starch, Maize, Technical, SLR, Fisher Chemical

Product Code. 10417080 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10417080 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10417080 Supplier Fisher Chemical Supplier No. S/7880/60
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1kg, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 5 to 7
CAS 9005-25-8
Physical Form Powder Solid
Molecular Formula C12H22O11
MDL Number MFCD00082026,MFCD00132834
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
Quantity 1 kg
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
PubChem CID 439341
ChEBI CHEBI:18167
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.