missing translation for 'onlineSavingsMsg'
Learn More

Starch, modified, insolubles 0.01% max

Product Code. 11374187
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374187 500 g 500g
11364187 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374187 Supplier Thermo Scientific Alfa Aesar Supplier No. 036673.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol

Starch, modified is used in the production of facial cleanser, oil controller, sunscreen cream, moisturizer, toners/astringents, facial moisturizer/treatment and anti-aging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00082026
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O

Specifications

Physical Form Powder
Solubility Information Instantly soluble in water
Quantity 500 g
Odor Odorless
Chemical Name or Material Starch, modified, Insolubles 0.01% max

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.