Learn More
Heidolph™ StarFish 5-Way Clamp
Product Code. 11731709 Shop All Heidolph Instruments Products
Click to view available options
Includes:
Silicone straps
Velcro straps
Unit Size:
Each
Product Code. 11731709
Brand: Heidolph™ 5058101000
Only null left
Add to basket
Add to basket
Hold glassware firmly in place on your StarFish magnetic stirrer workstation. Heidolph™ StarFish 5-Way Clamp features five telescopic arms which can be extended and locked in place to suit your needs.
  • For MR HEI Magnetic Stirrers and StarFish Workstations.
  • Holds glassware of virtually any size.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MR-HEI Magnetic Stirrers
Holds Glassware
Includes Velcro straps
Type Clamp
Voltage 100/115/220/230 V