missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StarGate pESG-IBA Vectors for Mammalian Expression

Product Code. 15623788 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pESG-IBA 102
pESG-IBA 103
pESG-IBA 104
pESG-IBA 105
pESG-IBA 123
pESG-IBA 142
pESG-IBA 143
pESG-IBA 145
pESG-IBA 162
pESG-IBA 164
pESG-IBA 167
pESG-IBA 168
pESG-IBA 23
pESG-IBA 25
pESG-IBA 3
pESG-IBA 33
pESG-IBA 35
pESG-IBA 43
pESG-IBA 45
pESG-IBA 5
pESG-IBA 62
pESG-IBA 63
pESG-IBA 64
pESG-IBA 65
pESG-IBA wt1
pESG-IBA wt2
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 15623788

Brand: IBA Lifesciences 54435001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for high-level constitutive expression of recombinant proteins in a wide range of Mammalian host cells through the CMV promoter, with replication in cell lines that are latently infected with SV40 large T antigen.

pESG-IBA mammalian expression vectors

pESG-IBA is a small expression vector recommended for transient expression of proteins in cell lines providing SV40 large T-antigen for extrachromosomal replication with SV40 ori.

  • NeoR for generation of stable cells lines.
  • Cytomegalovirus immediate-early (CMV) promoter: high level constitutive expression
  • SV40 ori: origin of replication for extrachromosomal replication driven by SV40 large T-antigen (as provided by HEK293T or COS-1, -3, -7 cells)
  • ColE1 + Amp: for propagation in E. coli
  • NeoR: neomycin resistance gene to generate stable cell lines
  • BM40: secretion signal to transfer the expressed protein into the medium (not for all vectors available)

Please note that cloning into pESG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pESG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pESG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.

Specifications

Format Liquid
For Use With (Equipment) StarGate™ System, Mammalian Host Cells
Inducing Agent SV40 large T antigen (e.g. COS1 or COS7)
Includes Neomycin resistance gene
pH 8
Promoter Human CMV Immediate-early
Product Type Acceptor Vector for mammalian expression
Storage Buffer 20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
Content And Storage 2°C to 8°C for frequent usage, -20°C for long-term storage
Protein Tag His Tag (6x) for N-terminal
Concentration 250 ng/μL
Quantity 5 μg
Vector pESG-IBA 35
Color Clear
For Use With (Application) Cloning Genes of Interest for Use in Functional Expression, Purification, Detection, Immobilization or Separation of Recombinant Proteins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.