missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences StarGate pESG-IBA Vectors for Mammalian Expression
GREENER_CHOICE

Product Code. 15663778 Shop All IBA Lifesciences Products
Change view
Click to view available options
Vector:
pESG-IBA 102
pESG-IBA 103
pESG-IBA 104
pESG-IBA 105
pESG-IBA 123
pESG-IBA 142
pESG-IBA 143
pESG-IBA 145
pESG-IBA 162
pESG-IBA 164
pESG-IBA 167
pESG-IBA 168
pESG-IBA 23
pESG-IBA 25
pESG-IBA 3
pESG-IBA 33
pESG-IBA 35
pESG-IBA 43
pESG-IBA 45
pESG-IBA 5
pESG-IBA 62
pESG-IBA 63
pESG-IBA 64
pESG-IBA 65
pESG-IBA wt1
pESG-IBA wt2
Unit Size:
5µg
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Vector unitSize
15663778 pESG-IBA wt2 5µg
15653778 pESG-IBA wt1 5µg
15673778 pESG-IBA 3 5µg
15683778 pESG-IBA 5 5µg
15693778 pESG-IBA 23 5µg
15603788 pESG-IBA 25 5µg
15613788 pESG-IBA 33 5µg
15623788 pESG-IBA 35 5µg
15633788 pESG-IBA 43 5µg
15643788 pESG-IBA 45 5µg
15653788 pESG-IBA 62 5µg
15663788 pESG-IBA 63 5µg
15673788 pESG-IBA 64 5µg
15683788 pESG-IBA 65 5µg
15693788 pESG-IBA 102 5µg
15603798 pESG-IBA 103 5µg
15613798 pESG-IBA 104 5µg
15623798 pESG-IBA 105 5µg
15633798 pESG-IBA 123 5µg
15643798 pESG-IBA 142 5µg
15653798 pESG-IBA 143 5µg
15663798 pESG-IBA 145 5µg
15673798 pESG-IBA 162 5µg
15683798 pESG-IBA 164 5µg
15693798 pESG-IBA 167 5µg
15603808 pESG-IBA 168 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15663778 Fornitore IBA Lifesciences N. del fornitore 54401001

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Designed for high-level constitutive expression of recombinant proteins in a wide range of Mammalian host cells through the CMV promoter, with replication in cell lines that are latently infected with SV40 large T antigen.

pESG-IBA mammalian expression vectors

pESG-IBA is a small expression vector recommended for transient expression of proteins in cell lines providing SV40 large T-antigen for extrachromosomal replication with SV40 ori.

  • NeoR for generation of stable cells lines.
  • Cytomegalovirus immediate-early (CMV) promoter: high level constitutive expression
  • SV40 ori: origin of replication for extrachromosomal replication driven by SV40 large T-antigen (as provided by HEK293T or COS-1, -3, -7 cells)
  • ColE1 + Amp: for propagation in E. coli
  • NeoR: neomycin resistance gene to generate stable cell lines
  • BM40: secretion signal to transfer the expressed protein into the medium (not for all vectors available)

Please note that cloning into pESG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pESG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pESG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.

Specifica

Format Liquid
For Use With (Equipment) StarGate™ System, Mammalian Host Cells
Inducing Agent SV40 large T antigen (e.g. COS1 or COS7)
Includes BM40 signal
pH 8
Promoter Human CMV Immediate-early
Product Type Acceptor Vector for mammalian expression
Storage Buffer 20 μL TE buffer: 10 mm Tris-HCl, 1 mm EDTA / 5 μg
Content And Storage 2°C to 8°C for frequent usage, -20°C for long-term storage
Protein Tag Untagged
Concentration 250 ng/μL
Quantity 5 μg
Vector pESG-IBA wt2
Color Clear
For Use With (Application) Cloning Genes of Interest for Use in Functional Expression, Purification, Detection, Immobilization or Separation of Recombinant Proteins
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.