Learn More
Abeba™ Static Control 31361 Shoes
Product Code. 15698797
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15698797 39 1 pair
15668797 36 1 pair
15678797 37 1 pair
15688797 38 1 pair
15608807 40 1 pair
15618807 41 1 pair
15628807 42 1 pair
15638807 43 1 pair
15648807 44 1 pair
15658807 45 1 pair
15668807 46 1 pair
15678807 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15698797 Supplier Abeba™ Supplier No. 31361/39
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative and Sanitized™ treated.
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather white
  • Composite toe cap
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Replaceable acc Wave insole (article no. 3553)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRA
Color White
Material Leather
Size 39
Sole Features Slip-resistant
Sole Material Rubber
Style 31361
Toe Material Composite