Learn More
Abeba™ Static Control 31366 Shoes
Product Code. 15648847
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15648847 36 1 pair
15658847 37 1 pair
15668847 38 1 pair
15678847 39 1 pair
15688847 40 1 pair
15698847 41 1 pair
15608857 42 1 pair
15618857 43 1 pair
15628857 44 1 pair
15638857 45 1 pair
15648857 46 1 pair
15658857 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15648847 Supplier Abeba™ Supplier No. 31366/36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative and Sanitized™ treated.
  • CE EN ISO 20345:2011 S1 SRA
  • Velours
  • Anthracite
  • Composite toe cap
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Replaceable acc Wave insole (article no. 3553)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRA
Color Anthracite
Material Velour
Size 36
Sole Features Slip-resistant
Sole Material Rubber
Style 31366
Toe Material Composite