Learn More
Abeba™ Static Control 31474 Shoes
Product Code. 15698927
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15698927 44 1 pair
15648927 39 1 pair
15658927 40 1 pair
15668927 41 1 pair
15678927 42 1 pair
15688927 43 1 pair
15608937 45 1 pair
15618937 46 1 pair
15628937 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15698927 Supplier Abeba™ Supplier No. 31474/44
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative and Sanitized™ treated.
  • CE EN ISO 20345:2011 S3 SRA
  • Smooth leather
  • Black in ATEX design
  • Composite toe cap
  • PU midsole
  • Metalfree penetration resistance
  • Slip resistance special rubber outsole sole
  • Replaceable acc Wave insole (article no. 3553)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S3 SRA
Color Black
Material Leather
Size 44
Sole Features Slip-resistant
Sole Material Rubber
Style 31474
Toe Material Composite