Learn More
Abeba™ Static Control 31475 Shoes
Description
- CE EN ISO 20345:2011 S3 SRA
- Smooth leather
- Black in ATEX design
- Composite toe cap
- PU midsole
- Metalfree penetration resistance
- Slip resistance special rubber outsole sole
- Replaceable acc Wave insole (article no. 3553)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20345:2011 S3 SRA |
| Color | Black |
| Material | Leather |
| Size | 39 |
| Sole Features | Slip-resistant |
| Sole Material | Rubber |
| Style | 31475 |
| Toe Material | Composite |