Learn More
Manutan Static control workbench, laminate, 1200mm x 600mm
Product Code. 12871488
Encompass_Preferred
Product Code. 12871488 Supplier Manutan Supplier No. A346510
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Very stable, fully welded cantilever workbenches. Stable, with four adjustable feet and earth grounding cord. Steel tube and sheet section framework to BS 6323. 14 and 16 swg.
  • Working height: 840mm
  • Dimensions (W x D): 1200mm x 600mm