Learn More
Ohaus™ Static Ionizer, ION-100A, EU
Product Code. 15466101 Shop All Ohaus Products
Change view
Click to view available options
Compatible with EX model electronic balances
Product Code.
15466101 Compatible with EX model electronic balances
1 options
Product Code. 15466101 Supplier Ohaus™ Supplier No. 30095929
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Neutralize the effects of static electricity, eliminate drift, and improve weighing precision.
  • Mitigates the effects of static electricity
  • Eliminates drift
  • Corrects the imbalance between positive and negative charge in an object
  • Improves accuracy and precision of weighing operations
  • Compatible with EX model electronic balances
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) EX Model Electronic Balance
For Use With (Application) Eliminates effects of static electricity on weighing, improves weighing accuracy
Languages Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Product Line Miscellaneous