missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cut Out Rubber Covered Rings

Product Code. 11765626 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Inner:
50 mm
75 mm
100 mm
125 mm
150 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
11765626 150 mm Each
11745626 100 mm Each
11735626 75 mm Each
11725626 50 mm Each
11755626 125 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11765626
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

With cut out section to allow easy removal of equipment.

  • With rubber covered rings
  • Stem length to the center of bosshead is approximately 60mm
  • Constructed in mild steel with welded joints
  • Zinc-plated

Specifications

Material Mild Steel
Diameter (Metric) Inner 150 mm
Product Type Stativring
Description Cut out section
Finish Zinc Plated, Rubber Coated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.