missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Staurosporine

Product Code. 18700957
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18700957 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18700957 Supplier Tocris Bioscience™ Supplier No. TB1285RMU/2
  • ONLINE EXCLUSIVE PRICE
    1955.00 €
    1848.00 € / 2mg
    Save 107.00 €
    5% Off
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Staurosporine synthesized to Ancillary Material Grade

TB1285-RMU is an Ancillary Material Grade version of product Staurosporine (Cat. No. 1285). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.

Find out more about our Ancillary Material Grade product range.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [9 S-(9α,10β,11β,13α)]-2,3,10,11,12,13-Hexahydro-10-methoxy-9-methyl-11-(methylamino)-9,13-epoxy-1 H,9 H-diindolo[1,2,3-gh:3 ',2 ',1 '-lm]pyrrolo[3,4-j][1,7 ]benzodiazonin-1-one
CAS 62996-74-1
Quantity 2 mg
Target Broad Spectrum Protein Kinase Inhibitors
Molecular Formula C28H26N4O3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.