missing translation for 'onlineSavingsMsg'
Learn More

Staurosporine, Fisher BioReagents

Product Code. 10647155
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Packaging:
Poly Micro Tube
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647155 1 mg Poly Micro Tube 1mg
10042804 100 μg Poly Micro Tube 100µg
10002854 500 μg Poly Micro Tube 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10647155 Supplier Fisher Bioreagents Supplier No. BP25411
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Staurosporine is a potent protein kinase inhibitor useful for studies on protein phosphorylation in the regulation of cellular functions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym staurosporine, kinome_3629
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Assay Percent Range > 98.0 %
Packaging Poly Micro Tube
Quantity 1 mg
Merck Index 15, 8928
Residual Solvents Pass Test
Formula Weight 466.5
Percent Purity >98.0%
Physical Form Solid
Chemical Name or Material Staurosporine, From Streptomyces sp.
compliance-icons
Product Identifier
  • Staurosporine
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.