missing translation for 'onlineSavingsMsg'
Learn More

Stavudine, TRC

Product Code. 30412449
Change view
Click to view available options
Quantity:
10 mg
25 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412449 25 mg
30415525 10 mg
30423514 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412449 Supplier Toronto Research Chemicals Supplier No. TRCS68525025MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3056-17-5
Molecular Formula C10 H12 N2 O4
Molecular Weight (g/mol) 224.21
Synonym Stavudine, 1-[(2R,5S)-5-(Hydroxymethyl)-2,5-dihydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Zidovudine Imp. A (EP), 1-(2,3-Dideoxy-β-D-glycero-pent-2-enofuranosyl)thymine, 3'-Deoxy-2'-thymidinene, 2',3'-Didehydro-3'-deoxythymidine, 3'-Deoxy-2',3'-didehydrothymidine, Avostav, BMY 27857, D 4T (nucleoside), NSC 163661, Sanilvudine, Staduvine, Stag, Stavir, Virostav, Zerit, d4T
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H12N2O4/c1-6-4-12(10(15)11-9(6)14)8-3-2-7(5-13)16-8/h2-4,7-8,13H,5H2,1H3,(H,11,14,15)/t7-,8+/m0/s1
Formula Weight 224.0797
Chemical Name or Material Stavudine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.