missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Stealth RNAi™ siRNA Negative Control Kit

Product Code. 10022642
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10022642 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10022642 Supplier Invitrogen™ Supplier No. 12935100
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stealth RNAi™ siRNA Negative Control Kit

Stealth RNAi™ siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi™ siRNA duplex versus background. These controls are:

  • Designed in 3 levels of GC content (Lo, Med, Hi) with 3 sequences available per level so researchers can match GC content to their experimental Stealth RNAi™ siRNA
  • Not homologous to anything in the vertebrate transcriptome
  • Tested not to induce stress response Stealth RNAi™ siRNA

    Negative Control Kit contains all three controls (Lo, Med, Hi GC content-excluding sequences #2 and #3) and each is also available separately. Get the ideal negative controls for your RNAi experiment so you can assess your results accurately.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Each individual control is supplied as 250 μl at 20 μM. The Stealth RNAi™ siRNA Negative Control Kit contains one each of all three negative controls (each 250 μl @ 20 μM). Store at -20°C. Products are guaranteed stable for 6 months when properly stored.
    Control Type Negative Control
    RNAi Type siRNA
    For Use With (Application) RNAi, Transfection
    Label or Dye Unconjugated
    Product Line Stealth RNAi™ siRNA
    Product Type siRNA
    Quantity 1 Kit

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.