missing translation for 'onlineSavingsMsg'
Learn More

Stearalkonium Chloride, TRC

Product Code. 30418949
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418949 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418949 Supplier Toronto Research Chemicals Supplier No. TRCS6857001G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stearalkonium Chloride
CAS 122-19-0
Purity Grade Notes HPLC
Molecular Formula C27 H50 N . Cl
Quantity 1 g
Synonym Benzenemethanaminium','N,N-dimethyl-N-octadecyl-','chloride (1:1), Ammonium','benzyldimethyloctadecyl-','chloride (8CI), Benzenemethanaminium','N,N-dimethyl-N-octadecyl-','chloride (9CI), Benzyldimethyloctadecylammonium chloride (6CI), 2B, 2B (onium compound), Algene SC 25, Alkaquat DMB-ST, Ammonyx 4, Ammonyx 4002, Ammonyx 490, Antistatic 1827, Arquad DM 18B90, Benzyldimethylstearylammonium chloride, Benzyloctadecyldimethylammonium chloride, Benzylstearyldimethylammonium chloride, Carsoquat SDQ 25, Carsoquat SDQ 85, Catinal OB 80E, Cation S (surfactant), Cation S 2-100, DC, DC (leveling agent), Dimethylbenzyloctadecylammonium chloride, Dimethyloctadecylbenzylammonium chloride, Dimethylstearylbenzylammonium chloride, Dodigen 1828, Incroquat S 85, Incroquat SDQ 25, Intexan SB 85, Intexsan SB 85, Larostat 905, Larostat HTS 905, Leveling agent 1827, Mackernium SDC 25, Mackernium SDC 85, Maquat SC 18, N,N-Dimethyl-N-benzyl-N-octadecylammonium chloride, N-Octadecyl-N-benzyl-N,N-dimethylammonium chloride, Nissan Cation S2-100, ODBAC 1827, Octadecylbenzyldimethylammonium chloride, Octadecyldimethylbenzylammonium chloride, Orthosan MB, Pentonium SD 75, Quaternol 1, SDQ 25, SDQ 85, Stearalkonium chloride, Stearylbenzyldimethyl ammonium chloride, Stearyldimethylbenzylammonium chloride, Stebac, Stedbac, Surfactant 1827, Swanol CA 1485, Tetranyl B 2MTH, Triton CG 400, Triton CG 500, Triton X 40, Triton X 400, Varisoft SDC, Varisoft SDC 85
InChI Formula InChI=1S/C27H50N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22-25-28(2,3)26-27-23-20-19-21-24-27;/h19-21,23-24H,4-18,22,25-26H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
IUPAC Name benzyl-dimethyl-octadecylazanium;chloride
Molecular Weight (g/mol) 424.15
Formula Weight 423.3632
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.