missing translation for 'onlineSavingsMsg'
Learn More

Stearic acid hydrazide, 97%

Product Code. 11385169
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385169 250 g 250g
11365169 10 g 10g
11375169 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11385169 Supplier Thermo Scientific Alfa Aesar Supplier No. A18291.30
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4130-54-5 | C18H38N2O | 298.515 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4130-54-5
Molecular Formula C18H38N2O
Molecular Weight (g/mol) 298.515
MDL Number MFCD00014764
InChI Key BYTFESSQUGDMQQ-UHFFFAOYSA-N
Synonym stearic acid hydrazide, stearic hydrazide, stearohydrazide, octadecanoic acid, hydrazide, stearoylhydrazine, stearic acid, hydrazide, unii-3fv1c68xop, 3fv1c68xop, octadecanohydrazide #, stearic acid hydrazone
PubChem CID 20088
IUPAC Name octadecanehydrazide
SMILES CCCCCCCCCCCCCCCCCC(=O)NN

Specifications

Melting Point 114°C to 116°C
Quantity 250 g
Formula Weight 298.51
Percent Purity 97%
Chemical Name or Material Stearic acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.