missing translation for 'onlineSavingsMsg'
Learn More

Stearoyl Ethanolamide, TRC

Product Code. 30422223
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422223 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422223 Supplier Toronto Research Chemicals Supplier No. TRCS686608500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stearoyl Ethanolamide
CAS 111-57-9
Molecular Formula C20H41NO2
Quantity 500 mg
Synonym N-(2-Hydroxyethyl)octadecanamide, AM 1105, Abriflo 65, Alkamide S 280, Aminon SM, Amisol SME, Ceramid, Clindrol 200MS, Comperlan HS, Cycloamide SM, Emcol 70, Loramine S 280 Lubsize K 12, Mackamide SMA, Marlamid M 18, Monamid 972, Monamid S, Monoethanolstearamide, N-(2-Hydroxyethyl)stearamide, N-Octadecanoylethanolamine, N-Stearoylethanolamine, NSC 3377, NSC 52619, Onyx Wax EL, Profan SME, Rewomid S 280, S 280, Stearamide MEA, Stearamide monoethanolamine, Stearic acid ethanolamide, Stearic acid monoethanolamide, Stearic ethanolamide, Stearic ethylolamide, Stearic monoethanolamide, Stearic monoethanolamine, Stearoylmonoethanolamide, Witcamide 70
InChI Formula InChI=1S/C20H41NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)21-18-19-22/h22H,2-19H2,1H3,(H,21,23)
SMILES CCCCCCCCCCCCCCCCCC(NCCO)=O
IUPAC Name N-(2-hydroxyethyl)octadecanamide
Molecular Weight (g/mol) 327.55
Formula Weight 327.3137
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.