missing translation for 'onlineSavingsMsg'
Learn More

Stearoyl L-alpha-Lysolecithin, TRC

Product Code. 30010346
Change view
Click to view available options
Quantity:
25 mg
250 mg
500 mg
Unit Size:
250mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010346 250 mg 250mg
30003694 500 mg 500mg
30006940 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010346 Supplier Toronto Research Chemicals Supplier No. TRCS686580250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19420-57-6
Molecular Formula C26H54NO7P
Molecular Weight (g/mol) 523.68
Synonym Stearoyl Lysophosphatidylcholine, 1-O-Stearoyl-2-lyso-sn-glycero-3-phosphocholine, 1-O-Stearoyl-sn-glycero-3-phosphocholine, 1-Octadecanoyl-sn-glycerol-3-phosphorylcholine, 1-Octadecanoyllysolecithin, 1-Stearoyl-sn-glycero-3-phosphocholine, 1-Stearoyl-sn-glycero-3-phosphorylcholine, 1-Stearoyl-sn-glycerol-3-phosphorylcholine, Coatsome MC 80H, Lysophosphatidylcholine C18:0, (7R)-4,7-Dihydroxy-N,N,N-trimethyl-10-oxo-3,5,9-trioxa-4-phosphaheptacosan-1-aminium 4-Oxide Inner Salt
IUPAC Name [(2R)-2-hydroxy-3-octadecanoyloxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCc1c(cccc1)N2C(=O)c3c(cccc3)N=C2C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C26H54NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-26(29)32-23-25(28)24-34-35(30,31)33-22-21-27(2,3)4/h25,28H,5-24H2,1-4H3/t25-/m1/s1
Formula Weight 523.3638
Chemical Name or Material Stearoyl L-alpha-Lysolecithin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.