missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 1 kg bag of 5.5 mm (.25 cal) stainless steel grinding beads
- Used to break hard, brittle samples specifically in applications including RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing
Specifications
Specifications
| Diameter (Metric) | 5.5 mm |
| Material | Stainless Steel |
| Packaging Quantity | 1 kg |
| Sample Type | Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores, Bacteria (Gram +/-), Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast, Sludge, Animal Tissue, Fixed Samples |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?