missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Steel Stand Bases

Steel Stand Bases with one adjustable foot.

Brand:  Bochem™ 5032

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10034930

  • 38.55 € / Each

Estimated Shipment: 19-04-2023
to see stock.
Add to basket

Description

Description

  • Thread M10
  • 1 foot adjustable
Specifications

Specifications

Steel
250 mm
Stand Base
6 mm
160 mm
1 Ea.
Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers