missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Steel Zincked Standard Retort Clamps with Cork

Brand:  Bochem™ 5482

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10035010

  • 39.00 € / Each
Estimated Shipment: 04-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1 Ea.
Zinc Plated Steel
180 mm
Stand Clamp
120 mm
120 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers