missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Stem-Cell Factor/c-Kit Inhibitor

Product Code. 15476729
Change view
Click to view available options
Quantity:
10mg
50mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476729 10mg 10mg
15486729 50mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476729 Supplier Thermo Scientific Chemicals Supplier No. J65114.MA
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Inhibits in vitro phosphorylation of c-Kit. Also inhibits SCF (stem cell factor)-induced c-kit phosphorylation in 501mel human melanoma cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 355.50
Quantity 10mg
Packaging Plastic bottle
Sensitivity Light sensitive
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.