missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Mesenchymal Progenitor Kit

Product Code. 16210521
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16210521 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16210521 Supplier STEMCELL Technologies Supplier No. 05240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Defined culture kit for derivation and expansion of mesenchymal progenitor cells

  • STEMdiff™ Mesenchymal Progenitor Kit is a defined culture kit consisting of animal component-free (ACF) induction medium, expansion medium, and attachment substrate
  • It is optimized for the derivation of cells with mesenchymal progenitor cell (MPC)-like properties from human embryonic stem (ES) cells or induced pluripotent stem (iPS) cells
  • This kit provides a complete workflow of defined reagents for derivation and expansion of human ES- or iPS-derived MPCs

Specifications

Cell Type Mesenchymal Cells, PSC-Derived
For Use With (Application) Cell Culture, Differentiation
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.