missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Midbrain Neuron Maturation Kit

Product Code. 16844286
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16844286 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16844286 Supplier STEMCELL Technologies Supplier No. 1000041
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maturation kit for generation of functional midbrain-type neurons from human ES and iPS cell-derived neuronal precursor cells

  • The STEMdiff™ Midbrain Neuron Maturation Kit (Catalog #100-0041) is used in conjunction with the STEMdiff™ Midbrain Neuron Differentiation Kit (Catalog #100-0038) to generate a mixed population of midbrain-type neurons from neural progenitor cells derived from human pluripotent stem cells
  • These media will produce a population of midbrain neurons (≥ 15% TH-positive dopaminergic neurons; ≥ 90% class III βtubulin-positive neurons; < 10% GFAP-positive astrocytes)
  • Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation

Specifications

Cell Type Dopaminergic Neurons, Neural Cells, PSC-Derived, Neurons
For Use With (Application) Cell Culture; Characterization; Differentiation; Functional Assay; Immunocytochemistry; Phenotyping
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.