missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ StemXVivo Mouse/Rat Osteogenic Supplement, 12.5 mL

Product Code. 18401870
Change view
Click to view available options
Quantity:
12.5 mL
Unit Size:
12.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18401870 12.5 mL 12.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401870 Supplier R&D Systems™ Supplier No. CCM009
  • ONLINE EXCLUSIVE PRICE
    262.94 €
    233.10 € / 12.5mL
    Save 29.84 €
    11% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Media supplement to induce the differentiation of mouse or rat MSCs into osteocytes

Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into osteocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.

  • Contains high quality differentiation factors to drive reproducible and efficient MSC osteogenesis
  • Is defined to reduce unwanted experimental variability
  • Has been developed and optimized using MSCs

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Supplement Media
Product Line R&D Systems
Quantity 12.5 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.