missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Steranios 2%

Product Code. 11794296
Click to view available options
Volume (Metric):
2 L
5 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11794296

Brand: Laboratoires Anios™ 382062

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High level disinfectant for medical devices surgical and medical equipment, endoscopy equipment and heat-sensitive equipment.

High level disinfectant:

  • Clear green solution.
  • Active against Helicobacter pylori.
  • 2% glutaraldehyde solution buffered at pH 6 (sodium citrate).
  • STERANIOS 2%NG and STERANIOS 2%ECS contain compounds
    limiting and controlling glutaraldehyde evaporation.
  • Wide compatibility with all kind of materials.
  • Shelf-life: 3 years from the production date.
  • Can be used in association with any kind of cleaning predisinfectant product.

Specifications

Product Type Steranios 2%
Composition Glutaraldehyde solution (2%) buffered at pH 6 in the presence a catalyst surface effect, dye and exc
For Use With (Application) For Utensils and Endoscopy Equipment
Volume (Metric) 2 L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • STERANIOS 2% 2L CAN
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : glutaraldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.