Learn More
Kern Sohn Stereo Zoom Microscope OZM
Product Code. 17369541
Encompass_Preferred
Change view
Click to view available options
Head Style:
Binocular
Trinocular
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Head Style Unit Size
17369541 Trinocular Each
17359541 Binocular Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17369541 Supplier Kern Sohn Supplier No. OZM 544
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
First-class optics and strong illumination combined with a high level of flexibility.
  • The KERN OZM-5 series is a range of excellent stereo zoom microscopes with above-average optical features.
  • The ergonomic shape allows a simple, effortless working over a period of several hours.
  • The extraordinarily strong and continuously dimmable 3W LED incident and transmitted illumination ensures a flexible and particularly good level of illumination for your sample.
  • With its large working distance, an extra large field of view and its brilliant resolution, the KERN OZM provides sharp, high-contrast, colour-true images.
  • The zoom objective offers you continuous magnification from 7×–45×.
  • The pillar stand is particularly flexible due to its variable and sturdy adjustment mechanism and therefore enables ergonomic working procedures.
  • A large selection of eyepieces, (universal) stands, a darkfield kit, external illumination units as well as auxiliary objectives and more are available as accessories
  • A protective dust cover, eye cups, as well as multi-lingual user instructions are included in the scope of delivery.
  • Scope of application: in vitro fertilisation, detection of parasites, zoology and botany, tissue preparation, section, quality control, Electronics and semiconductor industry, assembly and repair
  • Applications/Samples: Samples with focus on three-dimensional impression, zoom with variable magnification (depth, thickness), e.g. insects, seeds, circuit boards, components
Specifications
Voltage 230 V
Magnification Power 0.7X to 4.5X
Head Style Trinocular
Lighting Type LED