Learn More
Isofield™ Steridon Sterile Cleanroom Coverall with Hood - Grade A/B Compatible
BIOPHARMAGREENER_CHOICE
Product Code. 30178838
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 15
Pack of 20
Product Code. 30178838
Brand: Isofield™ 80111XS
Only null left
Add to basket
Add to basket
The tri-laminate Steridon coverall is aseptically folded and can be donned in 60 seconds or less, validated sterile via X-RAY, and tested against chemo to ASTM F 739. The anti-static, low linting, viral barrier provides essential protection in Grade A/B environments for all cleanroom activities.
  • Sterile Cleanroom coverall with hood
  • Latex-free, enclosed and elasticated face, wrists and ankles
  • Ergo-fit design at back and shoulders allowing greater range of motion
  • Aseptically presented fold
  • Color coded left and right inner tabs to facilitate ease of donning
  • Removable external tabs at zipper to ensure sterile field maintained
  • Validated for sterility by X-Ray
  • Sterility Assurance Level (SAL) 10-6
Technical and Documentation Compliance
  • COA, COC, COI Available for download online
  • QR Code on cartons allows for immediate Document retrieval
  • Permeation of Liquids and Gases through Protective Clothing under conditions of Continuous Contact ASTM F739-20
  • Resistance of Materials Used in Protective Clothing to Penetration by Synthetic Blood ISO 16603:2004
  • Viral penetration test (liquid challenge: Bacteriophage Phi-X174) ISO 16604:2004
  • Material resistance to wet bacterial Penetration ISO 22610
  • Resistance to penetration by biologically contaminated aerosols ISO 22611
  • Resistance to dry microbial penetration ISO 22612
  • Basis Weight ASTM D3776
  • Dry Mullen Burst Strength TAPPI T403
  • Grab Tensile Strength CD ASTM D5034
  • Grab Tensile Strength MD ASTM D5034
  • Trapezoid Tear MD ASTM D5733-95
  • Wet Peel CD (inner/outer) ASTM F904-98
  • Hydrostatic Head (with screen) AATCC 127
  • Moisture Vapor Transmission Rate (MVTR) ASTM E96 Procedure D
  • Static Decay MD/CD WSP 40.2
  • Sterility Validation EN ISO 11137 Part 2 : 2015
  • Particulate Cleanliness – Helmke Drum Test : Class 1
  • Surface Resistivity ASTM D257
Specifications
Product Type Cleanroom Coveralls
Size X-Small
Gender Unisex
Material Polyethylene, Non-woven Polypropylene Laminate
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Open
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Attached, Open Face
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Chemicals, Anti-static, Liquid, Particle
No. of Pockets None
Cleanroom Class Class 100 (ISO 5)