Learn More
Nitritex™ Sterile BioClean-D™ Disposable Laboratory Coat
Product Code. 15248298
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Size (Metric) Chest:
1066.8 to 1143 mm
1143 to 1219.2 mm
838.2 to 914.4 mm
914.4 to 990.6 mm
Unit Size:
Pack of 30
Product Code. 15248298
Brand: Nitritex™ SBDLCXL
Only null left
Add to basket
Add to basket
Single use offering excellent comfort and protection. The Sterile BioClean-D™ Disposable Laboratory Coat features stud fastenings, a rear vent and three pockets. Sterilized by gamma irradiation.
  • Press stud fastenings, open cuff & rear vent and three pockets
  • Exceptional comfort and protection
  • Single use disposable garment
  • Carefully researched, designed & constructed
  • Compatible for use in ISO Class 4 cleanrooms
  • Low linting & durable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lab Coat
Size X-Large
Material CleanTough Fabric
Color White
Cuff Style Open
Disposable Single-use
Sterility Sterile
No. of Pockets 3
Size (Metric) Chest 1066.8 to 1143 mm
Certifications/Compliance ISO 9001:2015, (EU)2016/425, EN ISO 13034:2005+A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2, ISO 11137-1:2006
Packaging Type One piece per sealed inner PE bag; one inner bag per sealed outer PE bag; 30 outer bags per lined carton (30 pieces)
Construction Spunbond Polypropylene