missing translation for 'onlineSavingsMsg'
Learn More

Technical Service Consultants™ Sterile Cotton Swabs

Product Code. 12719945
Click to view available options
Quantity:
1
Unit Size:
Pack of 500
This item is not returnable. View return policy

Product Code. 12719945

Brand: Technical Service Consultants™ TS6H

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use Technical Service Consultants™ Sterile Cotton Swabs to transport most fastidious and non-fastidious micro-organisms. They are also recommended for clinically significant samples.

  • Extensive range of swabs available with a choice of swab shafts including white birchwood, polypropylene (flexible) and polystyrene (breakable)
  • Tips formed from pure cotton, alginate and cotton
  • Gamma irradiated to ensure a SAL of at least 10-6

Specifications

Sterility Sterile
Handle Material Wood
Shelf Life 2 Years
Tip Material Cotton
Packaging In Tube
For Use With (Application) Hygiene and general purpose sampling
Color White
No. per Pack 500
Quantity 1
Product Type Swab
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.