missing translation for 'onlineSavingsMsg'
Learn More

Heinz Herenz Sterile Dry Swabs without Medium

Product Code. 15506809
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
15506809 Sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15506809 Supplier Heinz Herenz Supplier No. 1020002
  • ONLINE EXCLUSIVE PRICE
    41.36 €
    27.80 € / Pack of 100
    Save 13.56 €
    32% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dry Swabs without medium, sterile with plastic stopper.

We offer a wide range of sterile and nonsterile swabs:

  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.

Please note in the medical field:

  • Only suitable for single use
  • Use by medical personnel only

Specifications

Product Type Applicator Swab and Tube
Format Swab
Media Type Amies Agar
Sterility Sterile
Color White
For Use With (Equipment) Glass with 7.5 to 8.5 mm ID
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.