missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Sterile HydroPure SATWIPES™ System
BIOPHARMA

Product Code. 16341778 Shop All Contec Products
Change view
Click to view available options
Dimensions (L x W):
15 x 23 cm (6 x 9 in.)
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
16341778 15 x 23 cm (6 x 9 in.) Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16341778 Supplier Contec™ Supplier No. SSWC16HP
  • ONLINE EXCLUSIVE PRICE
    564.00 €
    478.85 € / Pack of 1000
    Save 85.15 €
    15% Off
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contec HydroPure is a blend of 6% hydrogen peroxide with purified water and Amplitude Theta polyester/cellulose wipes.

Efficacious against bacteria, fungi, moulds, yeasts and spores, Contec HydroPure leaves little-to-no residue making it ideal for use in critical areas.

Contec HydroPure is provided as a point-of-use SATWIPES system. Each double bagged pack contains a canister of 100 x 54 gsm Amplitude Theta polyester/cellulose dry wipes and a 500 mL capped bottle of Contec HydroPure.

Simply add 500 mL of HydroPure to the dry roll, 5 mins before use, and use the impregnated wipes within 7 days.

Contec HydroPure is 0.2 micron filtered, and filled in a Grade A environment. The sterile product is filtered into pre-irradiated components. The dry wipe canister is irradiated at no less than 25 kGy.

Features & Benefits

  • Impregnated wipes at point-of-use allow convenient and controlled application of disinfectant
  • Hydrogen Peroxide breaks down to water & oxygen leaving no residue
  • Sporicidal activity with very low residue so suitable for critical areas
  • Available sterile so ideal for a Grade A / B areas
  • Double bagged for easy transfer into cleanroom environment
  • Linear tear packaging makes each bag easy to open even when wearing gloves
TRUSTED_SUSTAINABILITY

Specifications

Product Type Dry Wipe
Length (English) 6 in.
Length (Metric) 15 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Polyester Cellulose
Dimensions (L x W) 15 x 23 cm (6 x 9 in.)
For Use With (Equipment) Contec HydroPure 6% hydrogen peroxide
Packaging Quantity 100/Canister, 10 Canisters/Cs.
Saturation Solution 6% Hydrogen Peroxide
compliance-icons
Product Identifier
  • CONTEC STERILE SATWIPES HYDROPURE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: hydrogen peroxide solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.