missing translation for 'onlineSavingsMsg'
Learn More

Technical Service Consultants™ Sterile Inoculation Needle

Product Code. 12719885
Change view
Click to view available options
Color:
Clear
White
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12719885 White Pack of 1000
12391309 Clear Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12719885 Supplier Technical Service Consultants™ Supplier No. TS/3C
  • ONLINE EXCLUSIVE PRICE
    224.00 €
    221.70 € / Pack of 1000
    Save 2.30 €
    1% Off
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use disposable Technical Service Consultants™ Sterile Inoculation Needle to save time compared to heat sterilization of re-usable needles. The needle is perfect for counting or isolating pathogens from heavy mixed cultures.

  • Tapered tip for improved bead security
  • Safe and secure transfer of cultures from freezer to culture plate
  • Eliminates need to balance beads with inoculating loop

Specifications

Disposable Single-use
Material Polystyrene
Sterility Sterile
Color White
Product Type Inoculating Loop
For Use With (Equipment) General laboratory use only
Quantity 1000/Pk.
For Use With (Application) General Laboratory use only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.