missing translation for 'onlineSavingsMsg'
Learn More

AGMA Sterile IPA 70% Disinfectant Spray

Product Code. 30066738
Change view
Click to view available options
Volume (Metric):
6 x 900 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30066738 6 x 900 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30066738 Supplier AGMA Supplier No. 105666
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Keep your Cleanroom environment clean and protected with Sterile IPA 70% in WFI disinfectant sprays and wipes.

The 0.2 μm filtered gamma irradiated Sterile Isopropanol 70% in WFI quality water is a proven industry standard biocide within pharmaceutical and hospital cleanrooms.

  • Efficacy independently validated.
  • WFI Quality Water – High Purity Water.
  • Ergonomic trigger with effective return mechanism.
  • Bag in Bottle to maintain sterility in use.
  • 13 week in-use Validation.
  • Fully traceability with supporting analysis.
  • Gamma irradiated for sterility.
  • 2-year unopened shelf life.
  • Double bagged (excluding 31824 and 192701) Trigger Sprays are filled under Grade A conditions
  • Available in sterile wet wipe, 500 mL, 1 L and 5 L fluid format.
  • Perfect for day-to-day disinfection across a range of surfaces including worktops, instruments, gloved hands and transferred items.

The Trigger sprays utilize The Bag-in-bottle technology to ensure in-use sterility, validated for 13 weeks. and are double bagged to safeguard product sterility during transfer and all packaging is Detox-dotted to give visual reassurance of successful irradiation.

Specifications

Product Type Isopropyl Alcohol
Form Liquid Spray
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Composition Sterile Isopropanol 70% v/v in WFI
Container Type Trigger Spray
Disposable Limited-use
Filtered Filtered
Flammable Yes
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Quantity 6 x 900 mL
Volume (Metric) 6 x 900 mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sterile IPA 70% in WFI 6
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol

Use biocides safely. Always read the label and product information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.