missing translation for 'onlineSavingsMsg'
Learn More

Chemsplash Sterile Irradiated OverBoot

Product Code. 30058264
Change view
Click to view available options
Cleanroom Class:
Class 100-1000 (ISO 5-6)
Unit Size:
200 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cleanroom Class Unit Size
30058264 Class 100-1000 (ISO 5-6) 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30058264 Supplier Chemsplash Supplier No. 5820.400.007
  • ONLINE EXCLUSIVE PRICE
    836.03 €
    774.30 € / 200 pairs
    Save 61.73 €
    7% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Supplied gamma sterile irradiated and double bagged, the Chemsplash Non Slip Overboot is elasticated at the calf and ankle and has a non slip white pvc sole.

Chemsplash Non Slip Overboot

  • Material: Chemsplash 55GSM Type PB 6B
  • Sterile Irradiated and Double Bagged
  • Elasticated Top and Ankles
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • White PVC Sole
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free

Description: Supplied GAMMA Sterile Irradiated and double bagged, the Chemsplash Non Slip Overboot is made from Chemsplash fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This overboot is elasticated at the calf and ankle and features a non-slip white PVC sole. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories are designed to protect the wearer from the environment or the environment from the wearer, ensuring maximum comfort and wearability.

Specifications

Size Universal
Material Microporous Fabric
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Elastic Top
Sole Type Anti-slip
Resistance Liquid
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100-1000 (ISO 5-6)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.