missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Sterile IPA
BIOPHARMA

Product Code. 15597171 Shop All Contec Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Container Type:
Capped Bottle
Trigger Spray Bottle
Quantity:
2 x 5 L
6 x 1 L
8 x 0.5 L
missing translation for 'unitSize':
2 bottles
6 bottles
8 bottles
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type Quantity missing translation for 'unitSize'
15597171 Trigger Spray Bottle 6 x 1 L 6 bottles
15892917 Trigger Spray Bottle 8 x 0.5 L 8 bottles
15537171 Capped Bottle 2 x 5 L 2 bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15597171 missing translation for 'mfr' Contec™ missing translation for 'supplierNo' SBT170IW
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provided sterile by gamma irradiated using a validated process at no less than 25 kGy. Contec™ Sterile IPA is an alcohol blend of 0.2 micron filtered, filled and bagged in a Grade C (ISO Class 7) cleanroom.

  • Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
  • GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
  • Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
TRUSTED_SUSTAINABILITY

Specifications

Product Type Ethyl Alcohol
Form Liquid Spray
pH Not Applicable
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Composition Isopropanol and WFI (Water for Injection)
Concentration 70% IPA in 30% WFI
Container Type Trigger Spray Bottle
Disposable Limited-use
Filtered Filtered
Flammable Yes
For Use With (Application) Sterilizes surfaces in cleanrooms
Quantity 6 x 1 L
Volume (English) 0.3 gal.
Volume (Metric) 1 L
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • CONTEC STERILE 70% IPA WITH WFI
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: PROPAN-2-OL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.