missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems
GREENER_CHOICE

Product Code. 11721263 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
20 μL
130 μL
250 μL
Packaging:
10 x 96 tips
50 x 96 tips
Fit for Purpose:
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Packaging Fit for Purpose unitSize
11721263 20 μL 50 x 96 tips Sterile and PCR Ready Pack of 4800
11701263 20 μL 10 x 96 tips Sterile Pack of 4800
11761263 130 μL 10 x 96 tips Sterile and PCR Ready Pack of 4800
11731263 250 μL 50 x 96 tips Sterile Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11721263 Supplier Thermo Scientific™ Supplier No. 918011
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Increase pipetting performance with these sterile pipette tips, which provide accurate and precise performance due to superior tip straightness and a low coefficient of variation.

Increase pipetting performance with Thermo ScientificTM Sterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Color Black
Format Rack
Tip Style Conductive
Packaging 50 x 96 tips
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile and PCR Ready
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.