missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Sterile Plastic Inoculation Loops (Volume: 1μL)

Product Code. 15782115
Change view
Click to view available options
Quantity:
50 Pouches of 20
800 Pouches of 20
Packaging Type:
Microseal Pouch
Microseal Pouch, Ecopack
Microseal bags
Sterility:
Aseptically Manufactured
Gamma-irradiated
Unit Size:
Pack of 1000
Pack of 16000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type Sterility unitSize
15782115 50 Pouches of 20 Microseal bags Aseptically Manufactured Pack of 1000
15762115 800 Pouches of 20 Microseal Pouch, Ecopack Aseptically Manufactured Pack of 16000
15772115 800 Pouches of 20 Microseal Pouch, Ecopack Gamma-irradiated Pack of 16000
15792115 50 Pouches of 20 Microseal Pouch Gamma-irradiated Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15782115 Supplier Microspec™ Supplier No. C4
In Stock
Add to basket
Add to basket

Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation.

  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Medium rigidity
  • Culture loops (Length: 200mm)
  • Smooth edges to avoid tearing and digging in agar
  • Volume: 1μL
  • Color: Blue

Specifications

Disposable Single-use
Volume 1 μL
Diameter (English) Inside 0.04 in.
Diameter (Metric) Inside 1 mm
Length (English) 7.87 in.
Length (Metric) 200 mm
Material High-impact Polystyrene
Sterility Aseptically Manufactured
Color Blue
Product Type Inoculating Product
Length (English) Handle 7.87 in.
Length (Metric) Handle 200 mm
Quantity 50 Pouches of 20
Packaging Type Microseal bags
Calibrated Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.