missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Sterile Plastic Inoculation Loops (Volume: 10μL)

Product Code. 15792105
Change view
Click to view available options
Quantity:
16000
50 Bags of 20
50 Pouches of 20
800 Pouches of 20
Packaging Type:
Microseal Pouch
Plastic Vial
Sterility:
Aseptically Manufactured
Gamma-irradiated
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 16000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type Sterility unitSize
15792105 50 Bags of 20 Microseal Pouch Aseptically Manufactured Pack of 1000
15702115 50 Pouches of 20 Microseal Pouch Gamma-irradiated Pack of 1000
15772105 16000 Plastic Vial Non-sterile Pack of 16000
15782105 800 Pouches of 20 Microseal Pouch Sterile Pack of 16000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15792105 Supplier Microspec™ Supplier No. A4
In Stock
Add to basket
Edge
Add to basket

Retrieves an inoculum from culture of microorganisms.

  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Medium rigidity
  • Culture loops (Length: 200mm)
  • Smooth edges to avoid tearing and digging in agar
  • Volume: 10μL
  • Color: Blue

Specifications

Disposable Single-use
Volume 10 μL
Diameter (English) Inside 0.18 in.
Diameter (Metric) Inside 4.5 mm
Length (English) 7.87 in.
Length (Metric) 200 mm
Material High-impact Polystyrene
Sterility Aseptically Manufactured
Color Blue
Product Type Plastic Culture Loops
Length (English) Handle 7.87 in.
Length (Metric) Handle 200 mm
Quantity 50 Bags of 20
Packaging Type Microseal Pouch
Calibrated Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.