missing translation for 'onlineSavingsMsg'
Learn More

Technical Service Consultants™ Sterile Polystyrene L-shaped Spreader

Product Code. 12301319
Change view
Click to view available options
Color:
Blue
Clear
Description:
Firm Plastic
Soft Plastic
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Description Unit Size
12301319 Clear Firm Plastic Pack of 1000
12769885 Blue Soft Plastic Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12301319 Supplier Technical Service Consultants™ Supplier No. TS/30C
  • ONLINE EXCLUSIVE PRICE
    173.00 €
    153.45 € / Pack of 1000
    Save 19.55 €
    11% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from polystyrene for strength and flexibility. Technical Service Consultants™ Sterile Polystyrene L-shaped Spreaders are ideal for spreading liquid or cultures uniformly over the surface of solid media.

  • Ideal for bacterial counts
  • General laboratory uses for sample throughput or protect
  • Sterilized by gamma irradiation; sterility guaranteed only while bag is sealed
  • Shelf life: 36 months from date of manufacture
  • Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container

Specifications

Description Firm Plastic
Color Clear
Quantity 1000/Pk.
Shape L-Shape
Material Polystyrene
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.