missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Reagent Reservoir

Product Code. 32214749 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
50 mL
55 mL
100 mL
Unit Size:
Pack of 100
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
32214749 100 mL Pack of 100
32214753 5 mL Pack of 50
32214752 10 mL Pack of 100
32214751 50 mL Pack of 100
32214744 55 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 32214749
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures

  • Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
  • Easy handling with tabs to prevent stacked reservoirs from sticking together
  • Manufactured and gamma irradiated in an ISO certified facility
  • Traceability certificate available upon request
  • Graduated 5, 10 mL

Specifications

Certifications/Compliance Certified DNase, RNase and Pyrogen Free
Color White
Capacity (Metric) 100 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) Pipetting
Packaging Individually Wrapped
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.