missing translation for 'onlineSavingsMsg'
Learn More

Seward™ Stomacher™ Model 3500 Sterile Bags

Product Code. 11854543
Change view
Click to view available options
Capacity (Metric):
400 mL to 3.5 L
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11854543 400 mL to 3.5 L Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11854543 Supplier Seward™ Supplier No. BA6042
Add to basket
Add to basket

The Stomacher™ Bag range has been designed to fulfill all applications from sample blending, straining and storage, including bags designed for extensive blending and small samples for life science applications.

  • Manufactured from high quality food grade virgin LDPE granule extruded to 65μm film thickness
  • Designed to withstand extensive blending without tears or perforations
  • Provides a low-cost option for sample preparation with no risk of cross-contamination between samples
  • All bags are irradiated sterile; certificate of irradiation is available upon request
  • Designed for blended homogenization of diluent samples for most applications

Specifications

Description Thickness of 65 microns
For Use With (Application) Blended homogenization of diluent samples
For Use With (Equipment) Stomacher 3500
Material LDPE
Quantity 250 (sachets of 50)
Dimensions (L x W) 380 x 510 mm
Capacity (Metric) 400 mL to 3.5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.