missing translation for 'onlineSavingsMsg'
Learn More

AGMA Sterile Zyceine Disinfectant

Product Code. p-200087502
Click to view available options
Quantity:
15 x 200 mL
3 x 1 L
Unit Size:
Pack of 15
Pack of 3
This item is not returnable. View return policy

Product Code. 30066742

Brand: AGMA 0064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protect your cleanrooms with a unique two-component sporicide that offers high microbial efficacy once activated.

Totally stable until activated, Zyceine has a proven fungicidal efficacy of greater than a 3-log reduction in 2 minutes, and a yeasticidal efficacy greater than a 4-log reduction in 2 minutes.

This solution has met the MHRA/NHS requirement for a sporicide for their 'wiping in' process which can achieve a 2 log reduction in 2mins. Available in a liquid pouch or high-quality wipes.

  • Fast acting Sporicid
  • Independently validated efficacy
  • WFI quality water – High Purity Water
  • Available with 28 x 38 cm wipes, allowing for large area coverage
  • Also available as 200 mL or 1 L fluid pouch, ideal for using with a mop system
  • Fully traceable with supporting analysis and certification
  • Gamma irradiation for Sterility
  • 2 year unopened helf life
  • 8 hour efficacy once activated
  • EN13704, EN1276, EN1650, EN14348

Specifications

Product Type Sterile Sporicide
Color-coded No
Form Liquid
pH 5 to 6
Scent Unscented
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Container Type Burstable Fluid Pouch
Disposable Limited-use
Filtered Filtered
Flammable No
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Quantity 3 x 1 L
Show More Show Less
Product Identifier
  • Sterile Zyceine in WFI
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium Hydroxide, Citric Acid Anhyrdous,Sodium Dichloroisocyanurate Anhydrous

Use biocides safely. Always read the label and product information before use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.